Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Specificity: ABS 052-121-02 cross-reacts with rat ghrelin, and is predicted to react with mouse, dog and cat based on sequence homology.
Reactivity: ABS 052-121-02 binds human ghrelin in solution. ABS 052-121-02 can be used as a detection antibody in a sandwich ELISA with capture antibody ABS 050-45-02.
NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/mL
GHRL encodes ghrelin-obestatin preproprotein, which generates ghrelin and obestatin. Ghrelin is an the ligand for the growth hormone secretagogue receptor type 1 and is involved in regulating growth hormone release. Ghrelin has an appeptite-stimulating effect and induces adiposity and stimulates gastric acid secretion. Obestatin may be the ligand for the orphan G protein-coupled receptor GPR39 and is involved in satiety and can have an appetite-reducing effect resulting in decreased food intake.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AMCBX1; Appetite-regulating hormone; Fip3p; ghrelin; ghrelin 27; ghrelin 28; ghrelin, growth hormone secretagogue receptor ligand; ghrelin/obestatin; ghrelin/obestatin preprohormone; Growth hormone secretagogue; Growth hormone-releasing peptide; IKK-gamma; In2c-preproghrelin; IP1; IP2; IPD2; motilin related peptide; Motilin-related peptide; OTTHUMP00000207794; OTTHUMP00000207795; OTTHUMP00000207796; OTTHUMP00000207798; prepro-appetite regulatory hormone; preproghrelin; proghrelin; Protein M46
Gene Aliases: 2210006E23Rik; Ghr; GHRL; m46; MTLRP; MTLRP-AP; MTLRPAP; UNQ524/PRO1066
UniProt ID: (Human) A8CF34, (Dog) Q9BEF8, (Cat) Q6BEG6, (Rat) Q9QYH7, (Mouse) Q9EQX0
Entrez Gene ID: (Human) 51738, (Dog) 403587, (Cat) 493844, (Rat) 59301, (Mouse) 58991
Molecular Function:
peptide hormone
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support